Sterilin® Pipettes, Dynalon
Supplier: Dynalon
Certificates
About this item
Disposable single-use Sterilin® pipettes are manufactured from crystal grade polystyrene for consistency and maximum clarity.
Pipettes feature easy-to-read crisp black printed graduations, enabling accurate use to ±1%. Sterility is guaranteed by gamma radiation at a license holding irradiation plant which follows the relevant recommendations in the DHSS Guide to Good manufacturing practice for sterile medical devices and surgical products. Sterilin® pipettes are tested for pyrogencity, and have a pyrogen level of less than 0.25 EU/ml.
Delivery: Pipettes are supplied in single wrap packaging.
Frequently Bought Together