Professional M3 Series Trimmer, Electron Microscopy Sciences
About this item
Our cutting head is made from die-cast alloy construction, which adds strength, maintains alignment. It’s molded to the shape of your hand with an exclusive Comfort Grip that makes repetitive work less fatiguing. Within the housing is a hollow ground tool-steel rotary blade that is self-sharpening for a long life of clean, precise cuts.
- Dual steel rails guide the cutting head with complete stability and precision for accurate, repeatable cuts
- Four nylon bearings in the head further ensure quiet, effortless cutting
- The extruded aluminum supports on each end lock in alignment
- A transparent paper guide automatically holds material in place without risk of damage
- An extruded aluminum alignment scale is factory-locked to be perfectly square to the blade edge
The alignment scale has measurements in inches and centimeters and accepts the Slide Guide for making repeatable cuts. The baseboard has a long lasting laminate finish that wipes clean in seconds. A durable, silk-screened registration grid, laid out in 1" increments, covers the baseboard for quick and easy measuring of any print or documents.
The cutting blade is completely covered by the blade housing – making the M3 Series an ideal choice for schools, offices, darkrooms, selfservice print stations, and high volume production labs. The base rests on non-slip rubber feet.
Made in the USA. Backed by a five year limited warranty.
2 Options Available Below
Product Details & Documents
Our cutting head is made from die-cast alloy construction, which adds strength, maintains alignment. It’s molded to the shape of your hand with an exclusive Comfort Grip that makes repetitive work less fatiguing. Within the housing is a hollow ground tool-steel rotary blade that is self-sharpening for a long life of clean, precise cuts.
- Dual steel rails guide the cutting head with complete stability and precision for accurate, repeatable cuts
- Four nylon bearings in the head further ensure quiet, effortless cutting
- The extruded aluminum supports on each end lock in alignment
- A transparent paper guide automatically holds material in place without risk of damage
- An extruded aluminum alignment scale is factory-locked to be perfectly square to the blade edge
The alignment scale has measurements in inches and centimeters and accepts the Slide Guide for making repeatable cuts. The baseboard has a long lasting laminate finish that wipes clean in seconds. A durable, silk-screened registration grid, laid out in 1" increments, covers the baseboard for quick and easy measuring of any print or documents.
The cutting blade is completely covered by the blade housing – making the M3 Series an ideal choice for schools, offices, darkrooms, selfservice print stations, and high volume production labs. The base rests on non-slip rubber feet.
Made in the USA. Backed by a five year limited warranty.
Product Family Options
Product Information
- Cutting capacityDimensionsWeightAvailabilityPrice
- 102098-120M3 Series Cutter, Model M3-24Supplier Number: 74885-24
- 102098-122M3 Series Cutter, Model M3-36Supplier Number: 74885-36