About this item
EZ-Accu Shot™ is a quantitative QC microorganism preparation designed for performing Growth Promotion Tests of culture media with ease.
EZ-Accu Shot™ kits include five vials of a single enumerated QC microorganism (one lyophilized pellet per vial), five vials of hydrating fluid (1.2 ml in each vial).
Each EZ-Accu Shot™ lyophilized QC microorganism preparation is designed to deliver 10 - 100 CFU per 0.1 ml of hydrated suspension as recommended by the USP/JP/Ph. Eur. With little to no preparation time required and up to eight hours of stability, EZ-Accu Shot™ adds efficiency and flexibility to QC laboratories’ busy schedules. Each 1.2 ml vial of hydrated suspension offers 10 inocula. For added convenience, a peel-off Certificate of Assay is included for quality control documentation.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Storage Temperature:2-8° C
- Stability:Up to eight hours of stability
Specifications
Related Information
Certificates
Search by Lot NumberInstructions for End User Registration with Microbiologics