- Protein/Peptide Type:Recombinant
- Source:E. coli
- Size:0.05 mg
- Protein Synonyms:XBP-1|XBP-1 XBP2|TREB5
- Protein/Peptide Name:Xbp1
- Purity:>95%
- Molecular Weight:25 kDa (Calculated)
- Formulation:Liquid
- Tested Applications:ELISA, Western Blot
- Cat. No.:10073-130
- Supplier no.:95-109
X box binding protein 1 (XBP-1) is a key protein in the mammalian unfolded protein response (UPR) that protects the cell against the stress of malfolded proteins in the endoplasmic reticulum (ER) (1). Upon sensing unfolded proteins, an ER transmembrane endonuclease and kinase termed IRE1p is activated and excises an intron from XBP-1 mRNA. The spliced XBP-1 mRNA results in a 371 amino acid protein (XBP-1s) which is then translocated to the nucleus where it binds to the regulatory elements of downstream genes. Together with other UPR transcription factors such as ATF6, XBP-1 stimulates the production of ER stress proteins including the ER resident protein chaperones glucose regulated protein (GRP) 78 and GRP94 (3,4).
This recombinant protein can be used for WB, ELISA, MS and neutralization assays. For research use only.
50mM Tris pH7.5, 150mM NaCl, 5 mM MgCl2.
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.