Order Entry
United States
ContactUsLinkComponent
Recombinant Puma Fragment (from E. coli)
Recombinant Puma Fragment (from E. coli)
Catalog # 10073-122
Supplier:  Prosci
CAS Number:  
undefined
Recombinant Puma Fragment (from E. coli)
Catalog # 10073-122
Supplier:  Prosci
Supplier Number:  95-102
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Size:
    0.05 mg
  • Protein Synonyms:
    JFY-1|PUMA JFY1|PUMA
  • Protein/Peptide Name:
    Puma Fragment
  • Purity:
    >95%
  • Molecular Weight:
    15 kDa (Calculated)
  • Formulation:
    Liquid
  • Tested Applications:
    ELISA, Western Blot
  • Cat. No.:
    10073-122
  • Supplier no.:
    95-102

Specifications

About this item

Apoptosis is related to many diseases and development. The p53 tumor-suppressor protein induces apoptosis through transcriptional activation of several genes. A novel p53 inducible pro-apoptotic gene was identified recently and designated PUMA (for p53 upregulated modulator of apoptosis) and bbc3 (for Bcl-2 binding component 3) in human and mouse (1-3). PUMA is one of the pro-apoptotic Bcl-2 family members including Bax and Noxa, which are also transcriptional targets of p53. The PUMA gene encodes two BH3 domain-containing proteins termed PUMA-α and PUMA-β (1). PUMA proteins bind Bcl-2, localize to the mitochondria, and induce cytochrome c release and apoptosis in response to p53. PUMA may be a direct mediator of p53-induced apoptosis. This PUMA recombinant protein is recognized by several ProSci PUMA monoclonal antibodies, but not the two polyclonal antibodies (PSI # 3041 and 3043).

This recombinant protein can be used for WB, ELISA, MS. For research use only.

1X PBS containing 0.1% SDS and 0.02% NaN.

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.