Order Entry
United States
ContactUsLinkComponent
Human Recombinant Calr (from E. coli)
Human Recombinant Calr (from E. coli)
Catalog # 10072-276
Supplier:  Prosci
CAS Number:  
undefined
Human Recombinant Calr (from E. coli)
Catalog # 10072-276
Supplier:  Prosci
Supplier Number:  XW-RP3028
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Protein Synonyms:
    Calreticulin|CRTC|CRT|cC1qR|SSA|CRP55|ERp60|HEL-S-99n|RO
  • Protein/Peptide Name:
    Calr
  • Purity:
    >90%
  • Molecular Weight:
    46 kDa (Calculated)
  • Formulation:
    Liquid
  • Tested Applications:
    ELISA, Western Blot, MS
  • Cat. No.:
    10072-276
  • Supplier no.:
    XW-RP3028

Specifications

About this item

Calreticulin is a multifunctional protein that acts as a major Ca(2+)-binding (storage) protein in the lumen of the endoplasmic reticulum. It is also found in the nucleus, suggesting that it may have a role in transcription regulation. Calreticulin binds to the synthetic peptide KLGFFKR, which is almost identical to an amino acid sequence in the DNA-binding domain of the superfamily of nuclear receptors. Calreticulin binds to antibodies in certain sera of systemic lupus and Sjogren patients which contain anti-Ro/SSA antibodies, it is highly conserved among species, and it is located in the endoplasmic and sarcoplasmic reticulum where it may bind calcium.

This recombinant protein can be used for WB, ELISA, MS. For research use only.

1X PBS containing 0.1% SDS and 0.02% NaN.

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.