Order Entry
United States
ContactUsLinkComponent
Human Recombinant GASP1 (from CHO Cells)
Human Recombinant GASP1 (from CHO Cells)
Catalog # 10073-076
Supplier:  Prosci
CAS Number:  
Human Recombinant GASP1 (from CHO Cells)
Catalog # 10073-076
Supplier:  Prosci
Supplier Number:  40-591
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    0.005 mg
  • Biological Activity:
    Determined by its ability to inhibit human Myostatin (GDF-8) activity in MCP-11 cells. The ED50 for this effect is 0.0025-0.0040 μg/mL in the presence of 5 ng/mL of human Myostatin (GDF-8).
  • Protein Synonyms:
    GASP-1|GASP1|KIAA0443|G-protein coupled receptor-associated sorting protein 1|GASP
  • Protein/Peptide Name:
    GASP1
  • Purity:
    >95%
  • Formulation:
    Lyophilized
  • Cat. No.:
    10073-076
  • Supplier no.:
    40-591

Specifications

About this item

Growth and differentiation factor-associated serum protein-1 (GASP-1) is a secreted inhibitory TGF-β binding protein that contains multiple protease inhibitor structural domains. It is expressed primarily in the ovary, testis, and brain, and can act as a potent soluble inhibitor of myostatin and GDF-11, but not Activin-A. The GASP-1 gene encodes a 571 amino acid protein that contains a 29 amino acid secretion signal sequence, and multiple identifiable structural features, including a WAP domain, a follistatin/Kazal domain, an immunoglobulin domain, two tandem Kunitz domains, and a netrin domain. Recombinant human GASP-1 is a 542 amino acid protein that migrates at an apparent molecular weight of approximately 55-66 kDa by SDS-PAGE analysis under non-reducing conditions.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.