Order Entry
United States
ContactUsLinkComponent
Human Recombinant IL34 (from HEK293 Cells)
Human Recombinant IL34 (from HEK293 Cells)
Catalog # 10073-032
Supplier:  Prosci
CAS Number:  
Human Recombinant IL34 (from HEK293 Cells)
Catalog # 10073-032
Supplier:  Prosci
Supplier Number:  40-560
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.002 mg
  • Biological Activity:
    Endotoxin level is less than 0.1 ng per μg (1EU/μg).
  • Protein Synonyms:
    C16orf77|Interleukin-34|IL-34
  • Protein/Peptide Name:
    IL34
  • Purity:
    >95%
  • Formulation:
    Lyophilized
  • Cat. No.:
    10073-032
  • Supplier no.:
    40-560

Specifications

About this item

IL-34 is a homodimeric cytokine that is expressed in a range of tissues including spleen, heart, brain, liver, kidney, lung, ovary, thymus, testis, small intestine, prostate and colon. IL-34 is a ligand for colony-stimulating factor-1 receptor (CSF1R), which also binds to CSF-1. It specifically binds CD14+ monocytes, promotes survival and proliferation of human peripheral blood monocytes, and stimulates macrophage colony formation by human bone marrow cells. IL-34, like human CSF-1 stimulates phosphorylation of MAPK1/ERK2 and MAPK3/ERK1. Recombinant human IL-34 is a 52.5 kDa homodimeric glycoprotein consisting of 460 amino acid residues, including a C-terminal His-Tag.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.