Order Entry
United States
ContactUsLinkComponent
Human Recombinant IL16 (from E. coli)
Human Recombinant IL16 (from E. coli)
Catalog # 10070-558
Supplier:  Prosci
CAS Number:  
Human Recombinant IL16 (from E. coli)
Catalog # 10070-558
Supplier:  Prosci
Supplier Number:  40-597
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.01 mg
  • Biological Activity:
    Determined by its ability to chemoattract human peripheral blood lymphocytes using a concentration range of 50.0-1000.0 ng/mL.
  • Protein Synonyms:
    NIL16|Lymphocyte chemoattractant factor|prIL-16|IL-16|PRIL16|Pro-interleukin-16|LCF
  • Protein/Peptide Name:
    IL16
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Tested Applications:
    Western Blot, ELISA
  • Cat. No.:
    10070-558
  • Supplier no.:
    40-597

Specifications

About this item

IL-16 was initially identified as a chemotactic cytokine (1), but is now known to possess a wide range of activities. Later studies have more fully characterized IL-16 as an immunomodulatory cytokine that contributes to the regulatory process of CD4+ T cell recruitment and activation at sites of inflammation in association with asthma and several autoimmune diseases (2). The precursor of IL-16 (pro-IL-16) is thought to be cleaved towards the C-terminal region by Caspase-3, releasing a 20 kDa active form that binds to and signals through CD4 (3,4). Besides acting as a chemotactic cytokine, IL-16 is thought to also be involved in the regulation of T cell proliferation (5) and multiple infectious, immune-mediated, and autoimmune inflammatory disorders including irritable bowel syndrome, systemic lupus erythematosus, and neurodegenerative disorders (6). At least two isoforms of IL-16 are known to exist; the longer isoform (also known as NIL-16) is detected only in neurons of the cerebellum and hippocampus (7). This recombinant protein fragment is derrived from this longer isoform.

This recombinant protein can be used for WB, ELISA, MS and neutralization assays.

1X PBS containing 0.1% SDS

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.