Order Entry
United States
ContactUsLinkComponent
Human Recombinant MIA2 (from E. coli)
Human Recombinant MIA2 (from E. coli)
Catalog # 10072-790
Supplier:  Prosci
CAS Number:  
Human Recombinant MIA2 (from E. coli)
Catalog # 10072-790
Supplier:  Prosci
Supplier Number:  40-227
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.005 mg
  • Environmentally Preferable:
  • Protein Synonyms:
    Melanoma inhibitory activity protein 2
  • Protein/Peptide Name:
    MIA2
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. No.:
    10072-790
  • Supplier no.:
    40-227

Specifications

About this item

MIA2 is a secreted cytokine and a member of the MIA/OTOR family. Members of this family which also includes MIA, OTOR, and TANGO share a Src homology-3 (SH3)-like domain. MIA2 is predominantly expressed in hepatocytes. Elevated levels of MIA2 may represent a clinically useful marker for diagnosis of hepatic disease activity and severity. Recombinant human MIA-2 is a 101 amino acid, 11.5 kDa protein comprising the MIA homologous N-terminal region of the full length MIA-2 protein.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.