Mouse Recombinant MIP3A (from E. coli)
0.005 mg
Catalog # 10072-124
Supplier: Prosci
Supplier #: 40-353
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
MIP-3 alpha is a CC chemokine that is expressed in the liver, lymph nodes, appendix, PBL and lung and signals through the CCR6 receptor. MIP-3 alpha is chemotactic towards lymphocytes and dendritic cells. Additionally, it promotes the adhesion of memory CD4+ T cells and inhibits colony formation of bone marrow myeloid immature progenitors. Recombinant murine MIP-3 alpha is a 7.9 kDa protein containing 70 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Documents
10072-124.pdf
Datasheet
Specifications
Specifications
- Size:0.005 mg
- Cat. No.:10072-124
- Biological Activity:Biological activity was determined by its ability to chemoattract murine CCR6 transfected HEK/293 cells using a concentration range of 0.1 - 10.0 ng/mL
- Protein/Peptide Type:Recombinant
- Formulation:Lyophilized
- Purity:>98%
- Protein Synonyms:MIP-3 alpha
- Protein/Peptide Name:MIP3A
- Species:Mouse
- Source:E. coli



