ContactUsLinkComponent
Modul-R™ Series Safety Goggles, 3M™ Company
Modul-R™ Series Safety Goggles, 3M™ Company
Catalog # 89095-252
Supplier:  3M
Modul-R™ Series Safety Goggles, 3M™ Company
Catalog # 89095-252
Supplier:  3M
Supplier Number:  7000030008
Specifications
  • Description:
    Modul-R™ series safety goggles, adjustable headband
  • Cat. No.:
    89095-252
  • Lens coating:
    Anti-fog
  • Lens tint:
    Clear
  • Supplier no.:
    40658-00000-10
Specifications
About this item
Modul-R™ Splash Goggles deliver reliable performance and protection in a snap.
  • Low-profile lens with an extended field of vision
  • Strap pivots for added adjustability
  • Splash shield has an adjustable venting system to help minimize fog and splash
Equipped with a splash shield and strap pivots for added adjustability, these goggles feature a low profile with an extended field of vision.
Certifications: Meets ANSI Z87.1-2003.