Order Entry
United States
ContactUsLinkComponent
Human Recombinant Fas Receptor (soluble) (from E. coli)
Human Recombinant Fas Receptor (soluble) (from E. coli)
Catalog # 10072-916
Supplier:  Prosci
CAS Number:  
Human Recombinant Fas Receptor (soluble) (from E. coli)
Catalog # 10072-916
Supplier:  Prosci
Supplier Number:  40-311
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.005 mg
  • Biological Activity:
    The ED50 was Biological activity was determined by its ability to inhibit the cytotoxicity of Jurkat cells is between 10 - 15 μg/mL in the presence of 2 ng/mL of hFasL.
  • Protein Synonyms:
    FASTM|Apo-1 antigen|APO-1|Tumor necrosis factor receptor superfamily member 6|ALPS1A|TNFRSF6|CD95|APT1|FAS1
  • Protein/Peptide Name:
    Fas Receptor (soluble)
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. No.:
    10072-916
  • Supplier no.:
    40-311

Specifications

About this item

Fas and Fas Ligand (FasL) belong to the TNF superfamily and are type I and type II transmembrane proteins, respectively. Binding of FasL to Fas triggers apoptosis in Fas-bearing cells. The mechanism of apoptosis involves recruitment of pro-caspase 8 through an adaptor molecule called FADD followed by processing of the pro-enzyme to active forms. These active caspases then cleave various cellular substrates leading to the eventual cell death. sFasR is capable of inhibiting FasL-induced apoptosis by acting as a decoy receptor that serves as a sink for FasL. The full length Fas (receptor) is a 319 amino acid type I transmembrane protein, which contains a 157 amino acid extracellular domain, a 17 amino acid transmembrane domain, and 145 amino acid cytoplasmic domain. Recombinant human soluble Fas (sFas Receptor) is a 157 amino acid polypeptide (17.6 kDa) corresponding to the TNFR homologous cysteine rich extracellular domain Fas.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.