To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.1 mg
- Protein Synonyms:PP1C|PP-1G|PPP1GProtein phosphatase 1C catalytic subunit
- Protein/Peptide Name:PPP1CC
- Purity:>95%
- Molecular Weight:37.1 kDa (Calculated)
- Tested Applications:ELISA, Western Blot, MS
- Cat. No.:10072-332
- Supplier no.:XW-RP3205
Specifications
About this item
Protein phosphatase 1 (PP1) is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. Involved in regulation of ionic conductances and long-term synaptic plasticity. May play an important role in dephosphorylating substrates such as the postsynaptic density-associated Ca(2+)/calmodulin dependent protein kinase II.
This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.002% NaN, 3mM NaCl, 2.5mM
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.