Human Recombinant Pole3 (from E. coli)
Catalog # 10072-328
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Protein Synonyms:CHARAC17|Arsenic-transactivated protein|DNA polymerase epsilon subunit 3|AsTP|p17|YBL1|CHRAC17
- Protein/Peptide Name:Pole3
- Purity:>95%
- Molecular Weight:16.9 kDa (Calculated)
- Tested Applications:ELISA, Western Blot, MS
- Cat. No.:10072-328
- Supplier no.:XW-RP3196
Specifications
About this item
POLE3 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging
This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.