Human Recombinant PFN2 (from E. coli)
Catalog # 10072-326
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.02 mg
- Protein Synonyms:Profilin-2|PFL|D3S1319E|Profilin II
- Protein/Peptide Name:PFN2
- Purity:>95%
- Molecular Weight:16.1 kDa (Calculated)
- Tested Applications:ELISA, Western Blot, MS
- Cat. No.:10072-326
- Supplier no.:XW-RP3189
Specifications
About this item
PFN2 protein is a ubiquitous actin monomer-binding protein belonging to the profilin family. It is thought to regulate actin polymerization in response to extracellular signals.
This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN, 10mM DTT
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.