Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant MTCBP-1 (from E. coli)
Catalog # 10072-314
Supplier:  Prosci
CAS Number:  
undefined
Human Recombinant MTCBP-1 (from E. coli)
Catalog # 10072-314
Supplier:  Prosci
Supplier Number:  XW-RP3154
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.1 mg
  • Protein Synonyms:
    MTCBP1|Ni-ARD|ARD|APL1|HMFT1638|Fe-ARD|2-dihydroxy-3-keto-5-methylthiopentene dioxygenase|Membrane-type 1 matrix metalloproteinase cytoplasmic tail-binding protein 1|mtnD|1|SIPL
  • Protein/Peptide Name:
    MTCBP-1
  • Purity:
    >95%
  • Molecular Weight:
    21.5 kDa (Calculated)
  • Tested Applications:
    ELISA, Western Blot, MS
  • Cat. No.:
    10072-314
  • Supplier no.:
    XW-RP3154

Specifications

About this item

MTCBP-1 is a new member of the Cupin superfamily with a role as an invasion suppressor down-regulated in tumors.

This recombinant protein can be used for WB, ELISA, MS.

10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.