Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant ATG7 (from E. coli)
Mouse Recombinant ATG7 (from E. coli)
Catalog # 10073-154
Supplier:  Prosci
CAS Number:  
undefined
Mouse Recombinant ATG7 (from E. coli)
Catalog # 10073-154
Supplier:  Prosci
Supplier Number:  95-122
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Protein Synonyms:
    Atg7l|ATG7|Gm21553|Apg7l|1810013K23Rik
  • Protein/Peptide Name:
    ATG7
  • Purity:
    >95%
  • Molecular Weight:
    30 kDa (Calculated)
  • Formulation:
    Liquid
  • Tested Applications:
    ELISA, Western Blot
  • Cat. No.:
    10073-154
  • Supplier no.:
    95-122

Specifications

About this item

Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components (1,2). This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1 (3). Another member of the autophagy family of proteins is APG7 which was identified in yeast as a ubiquitin-E1-like enzyme; this function is conserved in the mammalian homolog (4). In mammalian cells, APG7 is essential for autophagy conjugation systems, autophagosome formation, starvation-induced bulk degradation of proteins and organelles (5). It has been suggested that caspase-8 may alter APG7 levels and thus the APG7 program of autophagic cell death (6).

This recombinant protein can be used for WB, ELISA, MS and neutralization assays. For research use only.

20mM Tris, 0.5M NaCl, 10%glycerol, 250mM Imidazole, 5mM beta-mercaptoethanol

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.