ContactUsLinkComponent
Safety Needle Holder, Globe Scientific
Safety Needle Holder, Globe Scientific
Catalog # 10118-769
Supplier:  Globe Scientific
undefined
Safety Needle Holder, Globe Scientific
Catalog # 10118-769
Supplier:  Globe Scientific
Supplier Number:  1204
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Needle Holder with Safety Shield, Universal Fit
  • Cat. No.:
    10118-769
  • Supplier no.:
    1204
Specifications
About this item
This product is designed to protect blood collection personnel from accidental needle sticks.
  • Features a plastic shield that is used to cover the exposed needle after the collection of blood
  • An audible "click" indicates the needle is secured and ready for disposal
Fits both 13 and 16 mm blood collection tubes and all common needle units. This product complies with OSHA directives by preventing removal of the needle and re-use of holder.