Special Conductivity Cells, WTW
Catalog # 80086-736
Supplier: WTW
Specifications
- Description:2-Electrode Conductivity Cell LTA 1
- Cat. No.:80086-736
- Cell constant:1.0 cm⁻¹
- Supplier no.:301310
Specifications
About this item
Adapter cable required.