Anti-CK8 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Keratin 8 is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. The product of this gene typically dimerizes with keratin 18 to form an intermediate filament in simple single-layered epithelial cells. This protein plays a role in maintaining cellular structural integrity and also functions in signal transduction and cellular differentiation. Mutations in this gene cause cryptogenic cirrhosis.
Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100, Immunofluorescence: 1:100~1:200
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: KRT8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
10071-828.pdf
Datasheet
Specifications
- Size:0.1 mg
- Clonality:Polyclonal
- Molecular Weight:55 kDa
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific peptide.
- Form:Liquid
- Storage Buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- ImmunoFluorescence:Yes
- Host:Rabbit
- Concentration:1 mg/mL
- Cat. No.:10071-828
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:4 °C
- Antibody Type:Primary
- Antigen Symbol:KRT8
- Gene ID:P05787
- Immunogen:Keratin 8 (Ab-74) antibody was raised against a peptide sequence around aa.72~76 (L-L-S-P-L) derived from Human Keratin 8 (CK8).
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:KRT8
- Antigen Name:keratin 8
- Storage Temperature:Store antibody at –20 °C for up to one year.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



