Anti-STAT5A Rabbit Polyclonal Antibody
10068-806
: Prosci
- Antibody Type:Primary
- Antigen Name:signal transducer and activator of transcription 5A
- Antigen Symbol:STAT5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:P42229
- Antigen Synonyms:STAT5A
- Storage Buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Molecular Weight:90 kDa
- Storage Temperature:Store antibody at –20 °C for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:STAT5A (Ab-780) antibody was raised against a peptide sequence around aa. 778~782 (R-L-S-P-P) derived from Human STAT5A.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific peptide.
- Cat. No.:10068-806
- Supplier no.:79-290
Carries out a dual function: signal transduction and activation of transcription. Binds to the GAS element and activates PRL-induced transcription.
Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: STAT5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat