- Antibody Type:Primary
- Antigen Name:v-myc avian myelocytomatosis viral oncogene homolog
- Antigen Symbol:MYC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:P01106
- Antigen Synonyms:MYC
- Modification:Thr 58
- Storage Buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Molecular Weight:60 kDa
- Storage Temperature:Store antibody at –20 °C for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Myc (Phospho-Thr58) antibody was raised against a peptide sequence around phosphorylation site of threonine 58 (L-P-T (p) -P-P) derived from Human Myc.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-phosphopeptide.
- Cat. No.:10070-198
- Supplier no.:79-117
Participates in the regulation of gene transcription. Binds DNA in a non-specific manner, yet also specifically recognizes the core sequence 5'-CAC[GA]TG-3'. Seems to activate the transcription of growth-related genes.
Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100
Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-phosphopeptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: MYC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat