Specifications
- Antibody Type:Primary
- Antigen Name:PUUVsSgp1
- Antigen Symbol:PUUVsSgp1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Size:0.1 mg
- Form:Liquid
- Gene ID:P27313
- Antigen Synonyms:PUUVsSgp1
- Storage Buffer:Puumala virus nucleocapsid antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:N/A
- Storage Temperature:Puumala virus nucleocapsid antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Puumala virus nucleocapsid antibody was raised against a 19 amino acid peptide near the amino terminus of the Puumala virus nucleocapsid. The immunogen is located within amino acids 130 - 180 of Puumala Virus Nucleocapsid.
- Purification:Puumala virus nucelocapsid antibody is affinity chromatography purified via peptide column.
- Cat. No.:10062-074
- Supplier no.:7693
Specifications
About this item
Puumala virus (PUUV) is a rodent-borne hantavirus of the family Bunyaviridae, an enveloped, negative-sense RNA viruses with a tripartite genome that can cause hantavirus pulmonary syndrome (HPS) and is highly homologous to the protype hantavirus Hantaan virus. Like other hantaviruses, the PUUV glycoprotein is synthesized as a precursor that is posttranslationally processed into two glycoproteins G1 (Gn) and G2 (Gc). These glycoproteins interact with the PUUV nucleocapsid (NP) protein through their cytoplasmic tail, and this association has been suggested to be crucial to the binding of the ribonucleoprotein of the PUUV and the assembly of the virus particle.
Puumala virus nucleocapsid antibody can detect 10ng Puumala virus nucleocapsid peptide in ELISA at 1 ?g/ml.
Puumala virus nucelocapsid antibody is affinity chromatography purified via peptide column.
Puumala virus nucleocapsid antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: PUUVsSgp1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus