To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:PUUVsMgp1 glycoprotein polyprotein precursor
- Antigen Symbol:PUUVsMgp1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Size:0.1 mg
- Form:Liquid
- Gene ID:P27312
- Antigen Synonyms:PUUVsMgp1
- Storage Buffer:Puumala virus glycoprotein antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:N/A
- Storage Temperature:Puumala virus glycoprotein antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Puumala virus glycoprotein antibody was raised against a 19 amino acid peptide near the amino terminus of the Puumala virus glycoprotein. The immunogen is located within amino acids 90 - 140 of Puumala Virus Glycoprotein.
- Purification:Puumala virus glycoprotein antibody is affinity chromatography purified via peptide column.
- Cat. No.:10062-072
- Supplier no.:7691
Specifications
About this item
Puumala virus (PUUV) is a rodent-borne hantavirus of the family Bunyaviridae, an enveloped, negative-sense RNA viruses with a tripartite genome that can cause hantavirus pulmonary syndrome (HPS) and is highly homologous to the protype hantavirus Hantaan virus. Like other hantaviruses, the PUUV glycoprotein is synthesized as a precursor that is posttranslationally processed into two glycoproteins G1 (Gn) and G2 (Gc). These glycoproteins interact with the PUUV nucleocapsid (NP) protein through their cytoplasmic tail, and this association has been suggested to be crucial to the binding of the ribonucleoprotein of the PUUV and the assembly of the virus particle.
Puumala virus glycoprotein antibody can detect 10ng Puumala virus glycoprotein peptide in ELISA at 1 ?g/ml.
Puumala virus glycoprotein antibody is affinity chromatography purified via peptide column.
Puumala virus glycoprotein antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: PUUVsMgp1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus