- Antibody Type:Primary
- Antigen Name:SNVsMgp1 glycoprotein precursor
- Antigen Symbol:SNVsMgp1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Size:0.1 mg
- Form:Liquid
- Gene ID:Q89905
- Antigen Synonyms:SNVsMgp1
- Storage Buffer:Sin Nombre virus glycoprotein 2 antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:N/A
- Storage Temperature:Sin Nombre Glycoprotein 2 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Sin Nombre virus glycoprotein 2 antibody was raised against a 16 amino acid peptide near the center of the Sin Nombre virus glycoprotein. The immunogen is located within amino acids 670 - 720 of Sin Nombre Virus Glycoprotein 2.
- Purification:Sin Nombre virus glycoprotein 2 antibody is affinity chromatography purified via peptide column.
- Cat. No.:10062-064
- Supplier no.:7683
Sin Nombre virus (SNV) is a rodent-borne hantavirus of the family Bunyaviridae, an enveloped, negative-sense RNA viruses with a tripartite genome that can cause hantavirus pulmonary syndrome (HPS). Hantavirus glycoprotein precursor (GPC) is posttranslationally cleaved into two glycoproteins G1 (Gn) and G2 (Gc). While the G1 glycoprotein is is thought to be degraded by the host autophagy machinery, and this autophagic clearance is required for efficient virus repliaction (2), no such degradation was observed for the G2 glycoprotein.
Sin Nombre virus glycoprotein 2 antibody can detect 10ng Sin Nombre virus glycoprotein 2 peptide in ELISA at 1 ?g/ml.
Sin Nombre virus glycoprotein 2 antibody is affinity chromatography purified via peptide column.
Sin Nombre virus glycoprotein 2 antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: SNVsMgp1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus