To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:SNVsMgp1 glycoprotein precursor
- Antigen Symbol:SNVsMgp1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Size:0.1 mg
- Form:Liquid
- Gene ID:Q89905
- Antigen Synonyms:SNVsMgp1
- Storage Buffer:Sin Nombre virus glycoprotein 1 antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:N/A
- Storage Temperature:Sin Nombre Glycoprotein 1 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Sin Nombre virus glycoprotein 1 antibody was raised against a 19 amino acid peptide near the amino terminus of the Sin Nombre virus glycoprotein. The immunogen is located within amino acids 60 - 110 of Sin Nombre Virus Glycoprotein 1.
- Purification:Sin Nombre virus glycoprotein 1 antibody is affinity chromatography purified via peptide column.
- Cat. No.:10062-062
- Supplier no.:7681
Sin Nombre virus (SNV) is a rodent-borne hantavirus of the family Bunyaviridae, an enveloped, negative-sense RNA viruses with a tripartite genome that can cause hantavirus pulmonary syndrome (HPS). Hantavirus glycoprotein precursor (GPC) is posttranslationally cleaved into two glycoproteins G1 (Gn) and G2 (Gc). The G1 glycoprotein is is thought to be degraded by the host autophagy machinery, and this autophagic clearance is required for efficient virus repliaction.
Sin Nombre virus glycoprotein 1 antibody can detect 10ng Sin Nombre virus glycoprotein 1 peptide in ELISA at 1 ?g/ml.
Sin Nombre virus glycoprotein 1 antibody is affinity chromatography purified via peptide column.
Sin Nombre virus glycoprotein 1 antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: SNVsMgp1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus