Order Entry
United States
ContactUsLinkComponent
Anti-CBS Mouse Monoclonal Antibody [clone: [6B8]]
Anti-CBS Mouse Monoclonal Antibody [clone: [6B8]]
  10074-928
 :  Avantor
undefined
Anti-CBS Mouse Monoclonal Antibody [clone: [6B8]]
  10074-928
 :  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    cystathionine-beta-synthase
  • Antigen Symbol:
    CBS
  • Clonality:
    Monoclonal
  • Clone:
    [6B8]
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    0.05 mg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    P35520
  • Antigen Synonyms:
    CBS
  • Storage Buffer:
    PBS, pH 7.4
  • Storage Temperature:
    Aliquot and store CBS antibody at –20 °C or -80 °C. As with all antibodies avoid freeze/thaw cycles.
  • Shipping Temperature:
    4 °C
  • Immunogen:
    CBS (NP_000062, 1 a.a. ~ 101 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
  • Purification:
    Protein A Column
  • Cat. No.:
    10074-928
  • Supplier no.:
    51-894

 

 

CBS antibody can be used in Western Blot, immunohistochemistry starting at 10 ?g/mL, and flow cytometry starting at 0.5 µg/1E6 cells.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CBS
Clonality: Monoclonal
Clone: [6B8]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human