To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:histone cluster 1, H4a
- Antigen Symbol:HIST1H4A
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:0.05 mg
- Form:Liquid
- Gene ID:P62805
- Antigen Synonyms:HIST1H4A
- Storage Buffer:PBS.
- Storage Temperature:Histone H4 antibody may be stored at 4 °C for short-term only. As with all antibodies avoid freeze/thaw cycles. Store at –20 °C for long term. Aliquots are stable for at least 12 months
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:KLH-conjugated, synthetic peptide corresponding to amino acids 17-28 (RHR(dimeK) VLRDNIQGC) of human histone H4.
- Purification:Protein G Column
- Cat. No.:10077-160
- Supplier no.:51-635
Specifications
About this item
Histone H4 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.
PBS.
Histone H4 antibody may be stored at 4 °C for short-term only. As with all antibodies avoid freeze/thaw cycles. Store at -20 °C for long term. Aliquots are stable for at least 12 months
Type: Primary
Antigen: HIST1H4A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Rat