Anti-DEFB104A Goat Polyclonal Antibody
Catalog # 10115-036
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:defensin, beta 104A
- Antigen Symbol:DEFB104A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Size:0.1 mg
- Form:Liquid
- Gene ID:Q8WTQ1
- Antigen Synonyms:DEFB104A
- Storage Buffer:DEFB104A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping Temperature:4 °C
- Immunogen:DEFB104A antibody was raised against a 14 amino acid synthetic peptide near the internal region of DEFB104A (aa 37-50).
- Purification:Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:10115-036
- Supplier no.:42-871
Specifications
About this item
ELISA: Antibody detection limit dilution 1:128000.
DEFB104A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: DEFB104A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: