Anti-PROP1 Goat Polyclonal Antibody
Catalog # 10114-118
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:PROP paired-like homeobox 1
- Antigen Symbol:PROP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Size:0.1 mg
- Form:Liquid
- Gene ID:O75360
- Antigen Synonyms:PROP1
- Storage Buffer:PROP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping Temperature:4 °C
- Immunogen:PROP1 antibody was raised against a 14 amino acid synthetic peptide near the N-Terminus of PROP1.
- Purification:PROP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:10114-118
- Supplier no.:42-154
Specifications
About this item
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Preliminary experiments in human cortex lysates gave no specific signal but low background (at antibody concentration up to 1 ?g/mL).
PROP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: PROP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: