Anti-Listeria Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10069-584
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Listeria sp. Antibody (FITC)
- Antigen Symbol:Listeria
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Reactivity:Bacteria
- Western Blot:Yes
- Size:1 mL
- Form:Liquid
- Storage Buffer:Listeria sp. antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing bovine serum albumin (10 mg/ml) as a stabilizer and 0.1% sodium azide preservative.
- Storage Temperature:Listeria sp. antibody can be stored at –20 °C, stable for one year.
- Shipping Temperature:4 °C
- Immunogen:Listeria monocytogenes
- Purification:Purified IgG fraction of antiserum covalently coupled with high purity Isomer I of fluorescein isothiocyanate. Care is taken to insure complete removal of any free fluorescein from the final product.
- Cat. No.:10069-584
- Supplier no.:35-310
Specifications
About this item
Purified IgG fraction of antiserum covalently coupled with high purity Isomer I of fluorescein isothiocyanate. Care is taken to insure complete removal of any free fluorescein from the final product.
Listeria sp. antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing BSA (10 mg/ml) as a stabilizer and 0.1% sodium azide preservative.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Listeria sp. Antibody (FITC)
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria