Specifications
- Antibody Type:Primary
- Antigen Name:sortilin-related receptor, L(DLR class) A repeats containing
- Antigen Symbol:SORL1
- Clonality:Monoclonal
- Clone:3B6A9 / 3B6B11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mL
- Gene ID:Q92673
- Antigen Synonyms:SORL1
- Storage Buffer:Ascitic fluid containing 0.03% sodium azide.
- Storage Temperature:SORL1 monoclonal antibody can be stored at –20 °C, stable for one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping Temperature:4 °C
- Immunogen:Purified truncated recombinant SORL1-MBP expressed in E. Coli strain BL21 (DE3).
- Cat. No.:10070-678
- Supplier no.:32-217
Specifications
About this item
SORL1 (sortilin-related receptor, L A repeats containing) also known as sorting protein-related receptor containing LDLR class A (SorLA), is a Type I membrane protein that may be involved in cell-cell interaction. SorLA, a single transmembrane receptor, binds LDL and transports it into cells by endocytosis. SorLA is synthesized as a proreceptor which is processed to the mature form by a furin-like propeptidase. It can also bind to RAP (receptor-associated protein). SorLA is a multifunctional endocytis receptor important in lipoprotein and protease uptake. The N-terminal propeptide, which is removed, can be cleaved by furin or homologous proteases. Endogenous SorLA binds the neuropeptide head activator (HA) and is important for HA signaling and function. The gene encoding for the protein maps to chromosome 8p23.1. SorLA is expressed mainly in brain (cerebral cortex, cerebellum and the occipital pole), but can also be found in liver, spinal cord, kidney, testis and pancreas.
Western Blot:1:500 - 1:2,000
IHC(P):1:500 - 1:2,000
ELISA:Propose dilution 1:10,000.
Determining optimal working dilutions by titration test.
Ascitic fluid containing 0.03% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: SORL1
Clonality: Monoclonal
Clone: 3B6A9 / 3B6B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human