- Antibody Type:Primary
- Antigen Name:cyclin-dependent kinase inhibitor 2A
- Antigen Symbol:CDKN2A
- Clonality:Monoclonal
- Clone:1E12E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mL
- Gene ID:P42771
- Antigen Synonyms:CDKN2A
- Storage Buffer:Ascitic fluid containing 0.03% sodium azide.
- Storage Temperature:P16 (Mouse and Human) monoclonal antibody can be stored at –20 °C, stable for one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping Temperature:4 °C
- Immunogen:Ni-NTA purified truncated recombinant P16 expressed in E. Coli strain BL21 (DE3).
- Cat. No.:10070-652
- Supplier no.:32-203
The progression of cells through the cell cycle is regulated by a family of protein kinases known as cyclin-dependent kinases (Cdks). The sequential activation of individual members of this family and their consequent phosphorylation of critical substrates promotes orderly progression through the cell cycle. The cyclins function as differentially expressed positive regulators of Cdks. Negative regulators of the cycle include the p53-inducible 21 kDa WAF1/Cip1 protein designated p21, Kip1 p27 and p16. The complexes formed by Cdk4 and the D-type cyclins have been strongly implicated in the control of cell proliferation during the G1 phase. It has recently been shown that p16 binds to Cdk4 and inhibits the catalytic activity of the Cdk4/cyclin D complex. Moreover, the gene encoding p16 exhibits a high frequency of homozygous deletions and point mutations in established human tumor cell lines.
Western Blot:1:500 - 1:2,000
IHC(P):1:500 - 1:2,000
ELISA:Propose dilution 1:10,000.
Determining optimal working dilutions by titration test.
Ascitic fluid containing 0.03% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CDKN2A
Clonality: Monoclonal
Clone: 1E12E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat