- Antibody Type:Primary
- Antigen Name:LIM domain containing preferred translocation partner in lipoma
- Antigen Symbol:LPP
- Clonality:Monoclonal
- Clone:8B3A11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mL
- Gene ID:Q93052
- Antigen Synonyms:LPP
- Storage Buffer:Ascitic fluid containing 0.03% sodium azide.
- Storage Temperature:LPP monoclonal antibody can be stored at –20 °C, stable for one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping Temperature:4 °C
- Immunogen:Purified truncated recombinantLPP-His is expressed in E.Coli strain BL21 (DE3).
- Cat. No.:10070-614
- Supplier no.:32-187
LIM containing lipoma preferred partner, it belongs to the zyxin family. The Zyxin family of proteins contains five members, Ajuba, LIMD1, LPP, TRIP6 and Zyxin. LPP, an 80 kDa protein and a LIM domain-containing scaffolding protein contains three LIM domains at its carboxy terminus, which are preceded by a proline-rich pre-LIM region containing a number of protein interaction domains. The LPP localizes to sites of cell adhesion, such as focal adhesions and cell-cell contacts, and shuttles to the nucleus where it has transcriptional activation capacity.
Western Blot:1:500 - 1:2,000
IHC(P):1:500 - 1:2,000
IF:1:500 - 1:2,000
ELISA:Propose dilution 1:10,000.
Determining optimal working dilutions by titration test.
Ascitic fluid containing 0.03% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: LPP
Clonality: Monoclonal
Clone: 8B3A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human