Anti-GSG1 Rabbit Polyclonal Antibody
Catalog # 10110-958
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:germ cell associated 1
- Antigen Symbol:GSG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q6UXU4
- Antigen Synonyms:GSG1
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:36 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store GSG1 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human GSG1.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10110-958
- Supplier no.:30-948
Specifications
About this item
GSG1 belongs to the GSG1 family. GSG1 may cause the redistribution of PAPOLB from the cytosol to the endoplasmic reticulum.
GSG1 antibody can be used for detection of GSG1 by ELISA at 1:1562500. GSG1 antibody can be used for detection of GSG1 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: GSG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human