Specifications
- Antibody Type:Primary
- Antigen Name:phosphoglycerate mutase 2 (muscle)
- Antigen Symbol:PGAM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:P15259
- Antigen Synonyms:PGAM2
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:29 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store PGAM2 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human PGAM2.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10110-016
- Supplier no.:30-361
Specifications
About this item
PGAM2 is the interconversion of 3- and 2-phosphoglycerate with 2,3-bisphosphoglycerate as the primer of the reaction. It can also catalyze the reaction of EC 5.4.2.4 (synthase) and EC 3.1.3.13 (phosphatase), but with a reduced activity.
PGAM2 antibody can be used for detection of PGAM2 by ELISA at 1:62500. PGAM2 antibody can be used for detection of PGAM2 by western blot at 1.0 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: PGAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human