Specifications
- Antibody Type:Primary
- Antigen Name:insulin-like growth factor 2 mRNA binding protein 1
- Antigen Symbol:IGF2BP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q9NZI8
- Antigen Synonyms:IGF2BP1
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:63 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store IGF2BP1 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human IGF2BP1.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10108-120
- Supplier no.:29-384
Specifications
About this item
IGF2BP1 is a member of the IGF-II mRNA-binding protein (IMP) family. The protein contains four K homology domains and two RNA recognition motifs. It functions by binding to the 5' UTR of the insulin-like growth factor 2 (IGF2) mRNA and regulating IGF2 translation.This gene encodes a member of the IGF-II mRNA-binding protein (IMP) family. The protein encoded by this gene contains four K homology domains and two RNA recognition motifs. It functions by binding to the 5' UTR of the insulin-like growth factor 2 (IGF2) mRNA and regulating IGF2 translation.This gene encodes a member of the IGF-II mRNA-binding protein (IMP) family. The protein encoded by this gene contains four K homology domains and two RNA recognition motifs. It functions by binding to the 5' UTR of the insulin-like growth factor 2 (IGF2) mRNA and regulating IGF2 translation. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
IGF2BP1 antibody can be used for detection of IGF2BP1 by ELISA at 1:312500. IGF2BP1 antibody can be used for detection of IGF2BP1 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: IGF2BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat