Specifications
- Antibody Type:Primary
- Antigen Name:AT rich interactive domain 3B (BRIGHT-like)
- Antigen Symbol:ARID3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:O95443
- Antigen Synonyms:ARID3B
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:61 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store ARID3B antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human ARID3B.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10107-352
- Supplier no.:28-946
Specifications
About this item
ARID3B is a member of the ARID (AT-rich interaction domain) family of DNA-binding proteins. The protein is homologous with two proteins that bind to the retinoblastoma gene product, and also with the mouse Bright and Drosophila dead ringer proteins. Members of the ARID family have roles in embryonic patterning, cell lineage gene regulation, cell cycle control, transcriptional regulation and possibly in chromatin structure modification. This gene is a member of the ARID (AT-rich interaction domain) family of proteins which bind DNA. It is homologous with two proteins that bind to the retinoblastoma gene product and also with the mouse Bright and Drosophila dead ringer proteins. A pseudogene on chromosome 1p31 also exists for this gene. Other ARID family members have roles in embryonic patterning, cell lineage gene regulation, cell cycle control, transcriptional regulation and possibly in chromatin structure modification.This gene is a member of the ARID (AT-rich interaction domain) family of proteins which bind DNA. It is homologous with two proteins that bind to the retinoblastoma gene product and also with the mouse Bright and Drosophila dead ringer proteins. A pseudogene on chromosome 1p31 also exists for this gene. Other ARID family members have roles in embryonic patterning, cell lineage gene regulation, cell cycle control, transcriptional regulation and possibly in chromatin structure modification.
ARID3B antibody can be used for detection of ARID3B by ELISA at 1:312500. ARID3B antibody can be used for detection of ARID3B by western blot at 0.75 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: ARID3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog