To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:zinc finger protein 76
- Antigen Symbol:ZNF76
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 ul
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P36508
- Antigen Synonyms:ZNF76
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:62 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store ZNF76 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human ZNF76.
- Purification:Antibody is purified by protein A chromatography method.
- Cat. No.:10107-112
- Supplier no.:28-844
Specifications
About this item
ZNF76 belongs to the krueppel C2H2-type zinc-finger protein family. ZNF76 is a general transcription repressor targeting TATA-binding protein (TBP), through a process regulated by sumoylation. ZNF76 interacts with TBP through both its N and C termini, and both regions are required for ZNF76 to exert its inhibitory function on p53-mediated transactivation. As a TBP-interacting transcriptional modulator, ZNF76 is also regulated by alternative splicing.
ZNF76 antibody can be used for detection of ZNF76 by ELISA at 1:312500. ZNF76 antibody can be used for detection of ZNF76 by western blot at 1.25 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: ZNF76
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog