Specifications
- Antibody Type:Primary
- Antigen Name:tumor susceptibility gene 101
- Antigen Symbol:TSG101
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q61187
- Antigen Synonyms:TSG101
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:43 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store TSG101 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of mouse TSG101.
- Purification:Antibody is purified by protein A chromatography method.
- Cat. No.:10106-718
- Supplier no.:28-645
Specifications
About this item
TSG101 has a direct role in the control of growth and differentiation in primary epithelial cells. It is required for normal cell function of embryonic and adult tissues but that this gene is not a tumor suppressor for sporadic forms of breast cancer.
TSG101 antibody can be used for detection of TSG101 by ELISA at 1:1562500. TSG101 antibody can be used for detection of TSG101 by western blot at 1.25 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: TSG101
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat