Anti-NACC2 Rabbit Polyclonal Antibody
Catalog # 10101-024
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:NACC family member 2, BEN and BTB (POZ) domain containing
- Antigen Symbol:NACC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q96BF6
- Antigen Synonyms:NACC2
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:63 kDa, 52 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store BTBD14A antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human BTBD14A.
- Purification:Antibody is purified by protein A chromatography method.
- Cat. No.:10101-024
- Supplier no.:28-148
Specifications
About this item
cDNA sequence was generated by The National Institutes of Health Mammalian Gene Collection (MGC) Program.
BTBD14A antibody can be used for detection of BTBD14A by ELISA at 1:312500. BTBD14A antibody can be used for detection of BTBD14A by western blot at 0.625 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: NACC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog