Specifications
- Antibody Type:Primary
- Antigen Name:fructosamine 3 kinase related protein
- Antigen Symbol:FN3KRP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q9HA64
- Antigen Synonyms:FN3KRP
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:34 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store FN3KRP antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human FN3KRP.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10104-590
- Supplier no.:27-199
Specifications
About this item
FN3KRP phosphorylates psicosamines and ribulosamines, but not fructosamines, on the third carbon of the sugar moiety. Protein-bound psicosamine 3-phosphates and ribulosamine 3-phosphates are unstable and decompose under physiological conditions. Thus phosphorylation leads to deglycation.FN3KRP and FN3K (MIM 608425) protect proteins from nonenzymatic glycation by phosphorylating the modified amino acid. This phosphorylation destabilizes the sugar-amine linkage and leads to spontaneous decomposition (Conner et al., 2004 [PubMed 15381090]).
FN3KRP antibody can be used for detection of FN3KRP by ELISA at 1:1562500. FN3KRP antibody can be used for detection of FN3KRP by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: FN3KRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human