- Antibody Type:Primary
- Antigen Name:general transcription factor IIA, 1, 19/37kDa
- Antigen Symbol:GTF2A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:P52655
- Antigen Synonyms:GTF2A1
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:41 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store GTF2A1 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human GTF2A1.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10104-474
- Supplier no.:27-141
GTF2A1 belongs to the TFIIA subunit 1 family. It is a component of the transcription machinery of RNA polymerase II and plays an important role in transcriptional activation. This protein induces a conformational change within the TBP/TATA complex that enhances its stability under both in vitro and physiological salt conditions.
GTF2A1 antibody can be used for detection of GTF2A1 by ELISA at 1:62500. GTF2A1 antibody can be used for detection of GTF2A1 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: GTF2A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat