Multi-Parameter Meters, Handheld, Multi 3410 / 3420 / 3430, WTW
Supplier: Avantor
Certificates
About this item
Rugged multi-parameter meters for pH, ORP, DO (with optical sensor FDO®) and conductivity (available as 1-, 2- or 3-channel instrument)
- Brilliant high resolution TFT colour display with simultaneous display of 1, 2 or 3 parameters (colour coded)
- Tactile keypad for error-free operation and field proven, waterproof housing (IP 67)
- Continuous Measurement Control (CMC) for measuring in the optimum pH range
- Quality Sensor Control (QSC) benchmarks the current condition of the special IDS pH electrodes
- Digital processing and data transfer (GLP-compliant) via USB or mini USB interface
Ideal for portable field measurements.
Ordering information: Available in sets including sensors and accessories (field case, 4×NiMH rechargeable batteries (1,2 V), USB cable, power pack, CD-ROM). Additional sets are available on request. For more information please contact our Avantor Technical Office.
Frequently Bought Together