About this item
This antibody is specific to an 85 kDa protein known as XRCC1. X-ray repair cross complementing 1 (XRCC1) protein plays a role in excision repair of DNA after ionizing irradiation. XRCC1 binds to DNA ligase III through the C-terminal 96 amino acids and to DNA polymerase β through the N-terminal half. In testis XRCC1 is expressed at high levels.
Type:
Antigen: XRCC1
Clonality: Monoclonal
Clone: 33-2-5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10065-060
- 99990-608
- 10064-926
- Antigen Symbol:
- XRCC1
- XRCC1
- Antigen Name:
- X-ray Repair Complementing Defective Repair in Chinese cells 1
- X-ray Repair Complementing Defective Repair in Chinese cells 1
- Antigen Synonyms:
- complementing defective|X-ray-repair|X-ray repair cross-complementing protein 1|DNA repair protein XRCC1|repair in Chinese hamster
- X-ray-repair|complementing defective repair in Chinese hamster|X-ray repair cross-complementing protein 1|DNA repair protein XRCC1
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 33-2-5
- 33-2-5
- Host:
- Mouse
- Mouse
- Concentration:
- Concentrated
- Concentrated