Specifications
- Description:Mouse UnoVue HRP/DAB Detection System
- Cat. No.:10065-156
- Supplier no.:UM 100PD
- Sufficient for:100 Tests
Specifications
About this item
The Mouse UnoVue HRP/DAB Detection System is a polymer-based 1-step horseradish peroxidase-labeled detection system for immunohistochemistry. This kit can detect mouse IgG and IgM antibodies, with significantly lower background than biotin/avidin detection systems.
The Mouse UnoVue™ HRP/DAB Detection System can be used for manual staining or with any automated staining instrument. The working solution of stable DAB/Plus is stable for a week, offering convenience and cost savings due to less hazardous waste generation. This species-specific detection system is particularly useful for multiplex immunohistochemical staining assays.