ContactUsLinkComponent
Calgonate® Emergency First Aid Kit, Calgonate®
Calgonate® Emergency First Aid Kit, Calgonate®
Catalog # 102335-480
undefined
Calgonate® Emergency First Aid Kit, Calgonate®
Catalog # 102335-480
Supplier Number:  TS043-KIT01
Specifications
  • Description:
    Calgonate® Emergency First Aid Kit
  • Kit Contents:
    First Aid Instructions for HF Burns
    2 X Tubes of Calgonate® Gel (25g each)
    2 X Bottles of Calgonate® Emergency Eyewash (120mL Squirt Bottle)
    1 Pair of Medical Scissors
    1 Exposure Incident Report Sheet
    4 X (25/8 x 15/8") Oval Eyepad
    6 X 4x4" Gauze Pad
    2 X 3" Conforming Stretch Gauze Bandage
    2 X 2" Conforming Stretch Gauze Bandage
    2 X 59 mL Bottle of Antiseptic Spray
    2 X Ammonia Inhalant – Respiratory Stimulant
    2 X 5x9" Sterile Combine Pad
    1 X ½x180" Roll of Waterproof Adhesive Tape
    1 Cold Pack – Instant Cold Compress
    2 Pairs of Medical Gloves
    1 Pupil Gauge
    2 X 8x10" ABD Pad – Extra Absorbent
    6 X 3x3" Gauze Pad
  • Cat. No.:
    102335-480
  • Supplier no.:
    TS043-KIT01
Specifications
About this item
This convenient kit contains over 30 items for HF exposure response and general workplace first aid.