Calgonate® Emergency Hydrofluoric Acid Spill Kits, Calgonate®
Calgonate® Hydrofluoric Acid Bucket Spill Kits contain the personal protection equipment for users to safely neutralize and dispose of an Hydrofluoric acid spill
The 1-Bucket kits include a single 5-gallon bucket which contains personal protection equipment and supplies such as 1 tube of Calgonate® Calcium Gluconate gel (25g), 1 bottle of Calgonate® Eyewash (4oz.), and 4lbs. of neutralizer. The 2-Bucket kit consists of 2 bright 5-gallon buckets, containing personal protection equipment for 2 people and the products to neutralize and clean up hydrofluoric acid spills.
Frequently Bought Together