Order Entry
United States
ContactUsLinkComponent
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # RL705-703-125
undefined
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # RL705-703-125
Supplier Number:  705-703-125
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Donkey
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Goat
  • Western Blot:
    Yes
  • Size:
    500 µg
  • Epitope:
    Heavy and Light chain
  • Cross Adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Antigen Synonyms:
    F(ab')2 Anti-Goat IgG (H&L)
  • Antibody Fragment:
    F(ab')2
  • Storage Buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
  • Storage Temperature:
    –20 °C
  • Concentration:
    1.0 mg/ml by UV absorbance at 280 nm
  • Shipping Temperature:
    Ambient
  • Immunogen:
    Goat IgG whole molecule
  • Purification:
    purified by immunoaffinity chromatography
  • Cat. No.:
    RL705-703-125
  • Supplier no.:
    705-703-125

Specifications

About this item

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:500 - 1:5000;
Restore with deionized water (or equivalent); 500 µl
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Goat